Learn More
Methyl perfluorooctanoate, 97%, Thermo Scientific™
35.18 € - 108.54 €
Chemical Identifiers
CAS | 376-27-2 |
---|---|
Molecular Formula | C9H3F15O2 |
Molecular Weight (g/mol) | 428.097 |
MDL Number | MFCD00039244 |
InChI Key | XOCNYZFAMHDXJK-UHFFFAOYSA-N |
Synonym | methyl perfluorooctanoate, methyl pentadecafluorooctanoate, pentadecafluorooctanoic acid, methyl ester, methyl perfluorocaprylate, octanoic acid, pentadecafluoro-, methyl ester, perfluorooctanoic acid methyl ester, pentadecafluorooctanoic acid methyl ester, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, methyl ester, acmc-1cndk, methylpentadecafluorooctanoate |
PubChem CID | 67824 |
IUPAC Name | methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate |
SMILES | COC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
376-27-2 | |
428.097 | |
XOCNYZFAMHDXJK-UHFFFAOYSA-N | |
67824 | |
COC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
C9H3F15O2 | |
MFCD00039244 | |
methyl perfluorooctanoate, methyl pentadecafluorooctanoate, pentadecafluorooctanoic acid, methyl ester, methyl perfluorocaprylate, octanoic acid, pentadecafluoro-, methyl ester, perfluorooctanoic acid methyl ester, pentadecafluorooctanoic acid methyl ester, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, methyl ester, acmc-1cndk, methylpentadecafluorooctanoate | |
methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate |
Specifications
376-27-2 | |
158°C to 159°C | |
C9H3F15O2 | |
MFCD00039244 | |
methyl perfluorooctanoate, methyl pentadecafluorooctanoate, pentadecafluorooctanoic acid, methyl ester, methyl perfluorocaprylate, octanoic acid, pentadecafluoro-, methyl ester, perfluorooctanoic acid methyl ester, pentadecafluorooctanoic acid methyl ester, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, methyl ester, acmc-1cndk, methylpentadecafluorooctanoate | |
COC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F | |
428.097 | |
428.09 | |
Methyl perfluorooctanoate |
1.684 | |
65°C (149°F) | |
1.305 | |
1809817 | |
XOCNYZFAMHDXJK-UHFFFAOYSA-N | |
methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate | |
67824 | |
97% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'einecsNumber' : 206-808-1
missing translation for 'rtecsNumber' : RH0783200
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.