missing translation for 'onlineSavingsMsg'
Learn More

Methyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals

Methyl methacrylate, 99%, C5H8O2, CAS Number-80-62-6, 150-76-5, acryester m, diakon, methacrylic acid methyl ester, methyl 2-methyl-2-propenoate, methyl 2-methylpropenoate, methyl-methacrylat, methylmethacrylate, methyl methacrylate, methyl methylacrylate, pegalan, 1L

28.44 € - 253.00 €

Chemical Identifiers

CAS 80-62-6, 150-76-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008587
InChI Key VVQNEPGJFQJSBK-UHFFFAOYSA-N
Synonym methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
PubChem CID 6658
ChEBI CHEBI:34840
IUPAC Name methyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10562521
View Documents
Thermo Scientific Acros
127140250
25 mL Glass bottle
28.44 €
25mL
Please to purchase this item. Need a web account? Register with us today!
10071140
View Documents
Thermo Scientific Acros
127140010
1 L Glass bottle
48.44 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10153510
View Documents
Thermo Scientific Acros
127140025
2.5 L Glass bottle
94.17 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10791964
View Documents
Thermo Scientific Acros
127140100
10 L Plastic drum
253.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

80-62-6, 150-76-5
100.12
VVQNEPGJFQJSBK-UHFFFAOYSA-N
6658
methyl 2-methylprop-2-enoate
C5H8O2
MFCD00008587
methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
CHEBI:34840
CC(=C)C(=O)OC

Specifications

0.005 % max. (as methacrylic acid)
-48.0°C
0.9300g/mL
8°C
98.5% min. (GC)
H2C=C(CH3)CO2CH3
MFCD00008587
15, 6013
methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
VVQNEPGJFQJSBK-UHFFFAOYSA-N
methyl 2-methylprop-2-enoate
100.12
CHEBI:34840
100.12
Crystalline Powder or Crystals
80-62-6, 150-76-5
White to Yellow
100.0°C
Authentic
C5H8O2
1.4130 to 1.4150
02, II, 398
0.93
Solubility in water: 15.9g/L (20°C). Other solubilities: soluble in most common organic solvents such as me,tetrahydrofuran,esters,aromatic and chlorinated,ethanol,ether,acetone
CC(=C)C(=O)OC
10 to 110ppm MEHQ
6658
0.6 mPa.s (20°C)
99%
Methyl methacrylate, Stabilized
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Highly flammable liquid and vapor.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 201-297-1

Product Identifier
  • Methyl methacrylate

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.