missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl mercaptoacetate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Methyl thioglycolate was used in the preparation of 3-carbomethoxy-4- oxotetrahydrothiopyran, 2- and 4-carbomethoxy-3-oxotetrahydrothiophene. It is used to prepare methyl thioglycolate and aminoethanethiol conjugated gold nanorods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl thioglycolate was used in the preparation of 3-carbomethoxy-4- oxotetrahydrothiopyran, 2- and 4-carbomethoxy-3-oxotetrahydrothiophene. It is used to prepare methyl thioglycolate and aminoethanethiol conjugated gold nanorods.
Solubility
Soluble in water. (40 g/L at 20°C)
Notes
Air Sensitive. Store under dry inert gas. Incompatible with oxidizing agents and air.
Methyl thioglycolate was used in the preparation of 3-carbomethoxy-4- oxotetrahydrothiopyran, 2- and 4-carbomethoxy-3-oxotetrahydrothiophene. It is used to prepare methyl thioglycolate and aminoethanethiol conjugated gold nanorods.
Solubility
Soluble in water. (40 g/L at 20°C)
Notes
Air Sensitive. Store under dry inert gas. Incompatible with oxidizing agents and air.
Specifications
Specifications
Melting Point | -3°C |
Density | 1.163 |
Boiling Point | 148°C to 149°C |
Flash Point | 56°C (132°F) |
Odor | Stench |
Linear Formula | HSCH2COOCH3 |
Refractive Index | 1.466 |
Quantity | 100 g |
UN Number | UN1228 |
Beilstein | 506259 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'