missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azinphos Methyl, SPEX CertiPrep™
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Azinphos-Methyl | 86-50-0 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
CAS | 86-50-0, 67-64-1 |
Packaging | 2mL amber prescored glass ampule |
CAS Min % | 0.1, 99.9 |
Quantity | 1 mL |
CAS Max % | 0.1, 99.9 |
Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
Molecular Formula | C10H12N3O3PS2 |
InChI Key | CJJOSEISRRTUQB-UHFFFAOYSA-N |
SMILES | COP(=S)(OC)SCN1C(=O)C2=CC=CC=C2N=N1 |
Show More |
Safety and Handling
Product Identifier
- Methyl azinphos
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : acetone, azinphos-methyl
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'