missing translation for 'onlineSavingsMsg'
Learn More

Methyl azetidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15443456
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100202-39-9
Molecular Formula C5H10ClNO2
Molecular Weight (g/mol) 151.59
MDL Number MFCD01861758
InChI Key UOCWTLBPYROHEF-UHFFFAOYSA-N
Synonym methyl azetidine-3-carboxylate hydrochloride, azetidine-3-methyl carboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hydrochloride, 3-azetidinecarboxylic acid, methyl ester, hydrochloride, methyl azetidine-3-carboxylate hcl, methylazetidine-3-carboxylate hydrochloride, methyl 3-azetidinecarboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hcl, pubchem23516, acmc-209tka
PubChem CID 21100040
IUPAC Name methyl azetidine-3-carboxylate;hydrochloride
SMILES Cl.COC(=O)C1CNC1

Specifications

Boiling Point 187°C to 192°C
Flash Point 69°C (156°F)
Quantity 250 mg
Formula Weight 151.59
Percent Purity 95%
Chemical Name or Material Methyl azetidine-3-carboxylate hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'