missing translation for 'onlineSavingsMsg'
Learn More

Methyl Alcohol, Anhydrous, AR™, Macron Fine Chemicals™

Product Code. 15306121
2.5 L
Click to view available options
Quantity:
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Analytical grade solvent with a combination of value and purity for general laboratory and stockroom use.

AR™ grade solvents from Macron Fine Chemicals™ deliver reliable and consistent results to laboratories the world over. These solvents meet or exceed general laboratory standards for analytical grade products, helping ensure the integrity of results in applications.

  • Quality that ensures consistent results
  • Specifications that meet or exceed AR grade requirements
  • Produced in ISO certified facilities to ensure reliability

Specifications

Melting Point -97.6°C
CAS 67-56-1
Color Colorless
Acidity Passes test
Appearance Clear
Density 0.8g/mL
Boiling Point 64.7°C
Quantity 2.5 L
Flash Point 12°C
Physical Form Liquid
Residue after Evaporation ≤0.001%
Molecular Formula CH4O
Titratable Acid ≤0.0005meq/g
MDL Number MFCD00004595
Vapor Pressure 13.02 kPa
Water ≤0.1%
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
Solubility Information Soluble in water
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Formula Weight 32.04g/mol
Percent Purity ≥99.8%
Grade AR™
Chemical Name or Material Methyl Alcohol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl Alcohol, Anhydrous, AR™, Macron Fine Chemicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.