missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-acetylsalicylate, 98%, Thermo Scientific Chemicals

66.90 € - 191.00 €

Chemical Identifiers

CAS 16475-90-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00040924
InChI Key XLSMGNNWSRNTIQ-UHFFFAOYSA-N
Synonym methyl 5-acetylsalicylate, 5-acetylsalicylic acid methyl ester, 5-acetyl-2-hydroxybenzoic acid methyl ester, methyl 2-hydroxy-5-acetylbenzoate, benzoic acid, 5-acetyl-2-hydroxy-, methyl ester, 5-acetyl methyl salicylate, methyl 5-acethylsalicylate, pubchem22504, methyl5-acetylsalicylate, methyl-5-acetylsalicylate
PubChem CID 85444
IUPAC Name methyl 5-acetyl-2-hydroxybenzoate
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)OC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11469542
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10215.14
25 g
66.90 €
25g
Please to purchase this item. Need a web account? Register with us today!
11479542
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10215.22
100 g
191.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Methyl 5-acetylsalicylate is used in food flavorings and preservatives. It is useful in the relief of headache and muscle and joint aches It is also effective in reducing fever, inflammation, and swelling and thus has been used for treatment of rheumatoid arthritis, rheumatic fever, and mild infection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-acetylsalicylate is used in food flavorings and preservatives. It is useful in the relief of headache and muscle and joint aches It is also effective in reducing fever, inflammation, and swelling and thus has been used for treatment of rheumatoid arthritis, rheumatic fever, and mild infection.

Solubility
Soluble in methanol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications

Chemical Identifiers

16475-90-4
194.186
XLSMGNNWSRNTIQ-UHFFFAOYSA-N
85444
CC(=O)C1=CC(=C(C=C1)O)C(=O)OC
C10H10O4
MFCD00040924
methyl 5-acetylsalicylate, 5-acetylsalicylic acid methyl ester, 5-acetyl-2-hydroxybenzoic acid methyl ester, methyl 2-hydroxy-5-acetylbenzoate, benzoic acid, 5-acetyl-2-hydroxy-, methyl ester, 5-acetyl methyl salicylate, methyl 5-acethylsalicylate, pubchem22504, methyl5-acetylsalicylate, methyl-5-acetylsalicylate
methyl 5-acetyl-2-hydroxybenzoate

Specifications

16475-90-4
C10H10O4
2643824
Soluble in methanol.
CC(=O)C1=CC(=C(C=C1)O)C(=O)OC
194.186
194.19
Methyl 5-acetylsalicylate
58°C to 62°C
MFCD00040924
methyl 5-acetylsalicylate, 5-acetylsalicylic acid methyl ester, 5-acetyl-2-hydroxybenzoic acid methyl ester, methyl 2-hydroxy-5-acetylbenzoate, benzoic acid, 5-acetyl-2-hydroxy-, methyl ester, 5-acetyl methyl salicylate, methyl 5-acethylsalicylate, pubchem22504, methyl5-acetylsalicylate, methyl-5-acetylsalicylate
XLSMGNNWSRNTIQ-UHFFFAOYSA-N
methyl 5-acetyl-2-hydroxybenzoate
85444
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 240-532-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only