Learn More
Methyl 4-chlorocarbonylbenzoate, 98%, Thermo Scientific Chemicals
63.00 € - 900.00 €
Chemical Identifiers
CAS | 7377-26-6 |
---|---|
Molecular Formula | C9H7ClO3 |
Molecular Weight (g/mol) | 198.602 |
MDL Number | MFCD00059486 |
InChI Key | CVXXHXPNTZBZEL-UHFFFAOYSA-N |
Synonym | methyl 4-chlorocarbonyl benzoate, methyl 4-chlorocarbonylbenzoate, methyl 4-chloroformyl benzoate, terephthalic acid monomethyl ester chloride, methyl 4-carbonochloridoyl benzoate, methyl terephthaloyl chloride, benzoic acid, 4-chlorocarbonyl-, methyl ester, 4-chlorocarbonyl-benzoic acid methyl ester, 4-carbomethoxybenzoylchloride, 4-chlorocarbonylbenzoic acid methyl ester |
PubChem CID | 81850 |
IUPAC Name | methyl 4-carbonochloridoylbenzoate |
SMILES | COC(=O)C1=CC=C(C=C1)C(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15410908
|
Thermo Scientific Alfa Aesar
H54688.03 |
1 g |
63.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15420908
|
Thermo Scientific Alfa Aesar
H54688.06 |
5 g |
230.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15430908
|
Thermo Scientific Alfa Aesar
H54688.14 |
25 g | |||||||
Description
4-(Chlorocarbonyl)benzoic Acid Methyl Ester, is a building block, that can be used for the synthesis of various pharmaceutical compounds. It can be used for the preparation of benzophenone residues and also for the synthesis of novel novel bulky hydrophobic retinoids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Chlorocarbonyl)benzoic Acid Methyl Ester, is a building block, that can be used for the synthesis of various pharmaceutical compounds. It can be used for the preparation of benzophenone residues and also for the synthesis of novel novel bulky hydrophobic retinoids.
Solubility
Hydrolyzes in water.
Notes
Store at room temperature. Keep away from oxidizing agents and acids. Hygroscopic.
Chemical Identifiers
7377-26-6 | |
198.602 | |
CVXXHXPNTZBZEL-UHFFFAOYSA-N | |
81850 | |
COC(=O)C1=CC=C(C=C1)C(=O)Cl |
C9H7ClO3 | |
MFCD00059486 | |
methyl 4-chlorocarbonyl benzoate, methyl 4-chlorocarbonylbenzoate, methyl 4-chloroformyl benzoate, terephthalic acid monomethyl ester chloride, methyl 4-carbonochloridoyl benzoate, methyl terephthaloyl chloride, benzoic acid, 4-chlorocarbonyl-, methyl ester, 4-chlorocarbonyl-benzoic acid methyl ester, 4-carbomethoxybenzoylchloride, 4-chlorocarbonylbenzoic acid methyl ester | |
methyl 4-carbonochloridoylbenzoate |
Specifications
7377-26-6 | |
C9H7ClO3 | |
UN3261 | |
methyl 4-chlorocarbonyl benzoate, methyl 4-chlorocarbonylbenzoate, methyl 4-chloroformyl benzoate, terephthalic acid monomethyl ester chloride, methyl 4-carbonochloridoyl benzoate, methyl terephthaloyl chloride, benzoic acid, 4-chlorocarbonyl-, methyl ester, 4-chlorocarbonyl-benzoic acid methyl ester, 4-carbomethoxybenzoylchloride, 4-chlorocarbonylbenzoic acid methyl ester | |
CVXXHXPNTZBZEL-UHFFFAOYSA-N | |
methyl 4-carbonochloridoylbenzoate | |
81850 | |
≥98% |
52°C to 56°C | |
MFCD00059486 | |
Moisture sensitive | |
Hydrolyzes in water. | |
COC(=O)C1=CC=C(C=C1)C(=O)Cl | |
198.602 | |
198.6 | |
Methyl 4-chlorocarbonylbenzoate |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 230-937-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only