missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-trifluoromethylbenzoylacetate, 95%, Thermo Scientific™
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Specifications
Specifications
Color | Yellow |
Boiling Point | 94°C to 96°C (0.2 mmHg) |
Flash Point | >200°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Packaging | Glass bottle |
Refractive Index | 1.4864 to 1.4884 |
Quantity | 10 g |
Solubility Information | Solubility in water: neglegible in water |
Formula Weight | 246.18 |
Show More |
Safety and Handling
Product Identifier
- Methyl 3-trifluoromethylbenzoylacetate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction