missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-(2-bromoethoxy)benzoate, 97+%, Thermo Scientific™
58.70 € - 701.00 €
Chemical Identifiers
CAS | 59516-96-0 |
---|---|
Molecular Formula | C10H11BrO3 |
Molecular Weight (g/mol) | 259.10 |
MDL Number | MFCD06204041 |
InChI Key | CGGLKWTURIUCMN-UHFFFAOYSA-N |
Synonym | methyl 3-2-bromoethoxy benzoate, methyl 3-2-bromoethoxy benzenecarboxylate, benzoic acid,3-2-bromoethoxy-, methyl ester, methyl 3-2-bromoethoxy-benzoate, methyl 3-2-bromoethyloxy benzoate, 1-bromo-2-3'-carbomethoxyphenoxy ethane, 3-2-bromoethoxy benzoic acid methyl ester, 3-2-bromo-ethoxy-benzoic acid methyl ester |
PubChem CID | 10491483 |
IUPAC Name | methyl 3-(2-bromoethoxy)benzoate |
SMILES | COC(=O)C1=CC(OCCBr)=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15429127
|
Thermo Scientific Alfa Aesar
H52405.MD |
250 mg | |||||||
15409127
|
Thermo Scientific Alfa Aesar
H52405.03 |
1 g |
180.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15419127
|
Thermo Scientific Alfa Aesar
H52405.06 |
5 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsMethyl 3-(2-bromoethoxy)benzoate used in pharmaceutical research.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
59516-96-0 | |
259.10 | |
CGGLKWTURIUCMN-UHFFFAOYSA-N | |
10491483 | |
COC(=O)C1=CC(OCCBr)=CC=C1 |
C10H11BrO3 | |
MFCD06204041 | |
methyl 3-2-bromoethoxy benzoate, methyl 3-2-bromoethoxy benzenecarboxylate, benzoic acid,3-2-bromoethoxy-, methyl ester, methyl 3-2-bromoethoxy-benzoate, methyl 3-2-bromoethyloxy benzoate, 1-bromo-2-3'-carbomethoxyphenoxy ethane, 3-2-bromoethoxy benzoic acid methyl ester, 3-2-bromo-ethoxy-benzoic acid methyl ester | |
methyl 3-(2-bromoethoxy)benzoate |
Specifications
59516-96-0 | |
MFCD06204041 | |
CGGLKWTURIUCMN-UHFFFAOYSA-N | |
methyl 3-(2-bromoethoxy)benzoate | |
10491483 | |
≥97% |
C10H11BrO3 | |
methyl 3-2-bromoethoxy benzoate, methyl 3-2-bromoethoxy benzenecarboxylate, benzoic acid,3-2-bromoethoxy-, methyl ester, methyl 3-2-bromoethoxy-benzoate, methyl 3-2-bromoethyloxy benzoate, 1-bromo-2-3'-carbomethoxyphenoxy ethane, 3-2-bromoethoxy benzoic acid methyl ester, 3-2-bromo-ethoxy-benzoic acid methyl ester | |
COC(=O)C1=CC(OCCBr)=CC=C1 | |
259.10 | |
259.1 | |
Methyl 3-(2-bromoethoxy)benzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures