missing translation for 'onlineSavingsMsg'
Learn More

Methoxydimethyl(p-tolyl)silane, 94%, Thermo Scientific™

Product Code. 11357539
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51501-87-2
Molecular Formula C10H16OSi
Molecular Weight (g/mol) 180.32
MDL Number MFCD09266166
InChI Key GIQXGMLMIZAKLB-UHFFFAOYSA-N
Synonym methoxydimethyl p-tolyl silane, methoxy-dimethyl-4-methylphenyl silane, silane, methoxydimethyl 4-methylphenyl, dimethylmethoxy 4-methylphenyl silane, methoxydimethyl 4-methylphenyl silane, silane,methoxydimethyl 4-methylphenyl, amtsi072, dimethyl 4-methylphenyl methoxysilane, benzene,1-methoxydimethylsilyl-4-methyl
PubChem CID 142850
IUPAC Name methoxy-dimethyl-(4-methylphenyl)silane
SMILES CO[Si](C)(C)C1=CC=C(C)C=C1

Specifications

Quantity 5 g
Formula Weight 180.32
Percent Purity 94%
Sensitivity Moisture sensitive
Chemical Name or Material Methoxydimethyl(p-tolyl)silane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'