missing translation for 'onlineSavingsMsg'
Learn More

Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™

Product Code. 10163383
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Methanol (Peroxide-Free/Sequencing)

Methanol is used in staining and destaining protein electrophoresis gels, in HPLC, and in other biological applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Absorbance 0.025 max. at 254nm, 0.15 max. at 230nm, 0.3 max. at 220nm, 1 max. at 205nm
Color Colorless
Density 0.791g/cm³
Boiling Point 64.7°C
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.326 to 1.330 (at 25°C)
Assay Percent Range ≥99.9 %
Residue after Evaporation 3ppm max.
Titratable Acid 0.0003mEq/g max.
Linear Formula CH3OH
Titratable Base 0.0002mEq/g max.
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55 cP at 20°C
Water 0.1% max.
Merck Index 15, 6029
Formula Weight 32.04
Percent Purity ≥99.9%
Grade Peroxides: ≤0001%
Chemical Name or Material Methanol, Peroxide-free
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P240-Ground/bond container and receiving equipment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'