Learn More
Methanol-d4, for NMR, 99.5% atom D, Thermo Scientific Chemicals
95.78 € - 426.12 €
Chemical Identifiers
CAS | 811-98-3 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 36.066 |
MDL Number | MFCD00044637 |
InChI Key | OKKJLVBELUTLKV-MZCSYVLQSA-N |
Synonym | methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d |
PubChem CID | 71568 |
IUPAC Name | trideuterio(deuteriooxy)methane |
SMILES | CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10397770
|
Thermo Scientific Acros
166350050 |
5 mL | Glass Bottle |
95.78 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10635222
|
Thermo Scientific Acros
166350100 |
10 mL | Glass Bottle |
154.77 €
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10409080
|
Thermo Scientific Acros
166350500 |
50 mL | Glass Bottle |
426.12 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
811-98-3 | |
36.066 | |
OKKJLVBELUTLKV-MZCSYVLQSA-N | |
71568 | |
CO |
CH4O | |
MFCD00044637 | |
methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d | |
trideuterio(deuteriooxy)methane |
Specifications
Liquid | |
0.8880g/mL | |
12°C | |
128mbar at 20°C | |
811-98-3 | |
65.0°C | |
CD3OD | |
01, III, 1187 | |
OKKJLVBELUTLKV-MZCSYVLQSA-N | |
trideuterio(deuteriooxy)methane | |
71568 | |
NMR |
Colorless | |
99.4% min. | |
1.3262 | |
0.06% max. (Coulometric) | |
-99.0°C | |
CH4O | |
MFCD00044637 | |
methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d | |
CO | |
36.066 | |
36.06 | |
Methanol-d4, 99.5 atom % D |
Safety and Handling
GHS H Statement:
Causes damage to organs.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 212-378-6
missing translation for 'tsca' : TSCA
RUO – Research Use Only