missing translation for 'onlineSavingsMsg'
Learn More

Methanol, BAKER ULTRA RESI-ANALYZED, J.T. Baker™

Product Code. 10284591
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Gives superior performance with minimal risk of contaminants that can limit accuracy and productivity. Methanol, BAKER ULTRA RESI-ANALYZED, J.T. Baker™ is an ultra-pure solvent for organic residue analysis that maximizes an instrument’s sensitivity and detection power.

BAKER ULTRA RESI-ANALYZED solvents were developed for organic residue extraction/concentration procedures. These solvents are fully characterized and lot controlled by ECD, FID, or other method-specific detectors to deliver the highest level of purity and lot-to-lot consistency.

  • Unique stabilizer systems provide unmatched product stability and interference-free results when chemical requires
  • Optimized distillation targeted to remove reactive impurities, production of higher assays and narrow solvent fronts
  • Performance tested to purity levels below the Lower Limits of Quantitation (LLQ) for trace analyte detection
  • Performance is guaranteed through the expiration period stated on the label
  • Suitable for exacting EPA sample cleanup protocols

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Density 0.79g/cm3
Boiling Point 64.7°C
Quantity 2.5 L
Explosion Limit 5.5 - 44 Vol %
Flash Point 11°C
Ignition Point 455°C
Odor Alcohol-like
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 1 ppm
Titratable Acid Max. 0.3μeq/g
Linear Formula CH3OH
Titratable Base Max. 0.1μeq/g
Water Max. 0.08%
Solubility Information Fully miscible in water
Formula Weight 32.04g/mol
Percent Purity Min. 99.8%
Grade For Organic Residue Analysis
Chemical Name or Material Methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.