missing translation for 'onlineSavingsMsg'
Learn More

Methanol, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 15538464
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Exceeds ACS Specifications

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Hydroxy methane , Methyl alcohol , Carbinol , Wood alcohol
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Max. 10 (APHA)
Appearance Clear
Density 0.7918 g/cm³ (20°C)
Boiling Point 64.6°C (1013 hPa)
Quantity 2.5 L
Flash Point 11°C
Packaging Plastic Bottle
Assay Percent Range min. 99.8 % (by GC, corrected for water)
Residue after Evaporation max. 0.001 %
Water max. 0.05 %
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.