Learn More
Methanol, 99.8+%, ACS reagent, Thermo Scientific Chemicals
26.03 € - 452.25 €
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10607221
|
Thermo Scientific Acros
423955000 |
500 mL | EcoSafPak™ Glass Bottle |
26.03 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10107660
|
Thermo Scientific Acros
423950010 |
1 L | EcoSafPak™ Glass Bottle |
38.79 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10073023
|
Thermo Scientific Acros
423950025 |
2.5 L | Glass Bottle |
72.26 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10668323
|
Thermo Scientific Acros
423950250 |
25 L | Metal Drum |
452.25 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
-98.0°C | |
0.7910g/mL | |
12°C | |
1.3280 to 1.3300 | |
0.001% max. | |
CH4O | |
CH3OH | |
MFCD00004595 | |
0.55 mPa.s (20°C) | |
0.1% max. (K.F.) | |
15, 6029 | |
Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents | |
CO | |
32.04 | |
CHEBI:17790 | |
≥99.8% | |
Methanol |
67-56-1 | |
64.7°C | |
Liquid | |
99.8+% | |
0.791 | |
0.0003 meq/g max. | |
0.0002 meq/g max. | |
Vapor Pressure: 128hPa at 20°C | |
01, IV, 1227 | |
11,322; 13,337 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 | |
ACS Reagent |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Methanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P330-Rinse mouth.
- P363-Wash contaminated clothing before reuse.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methanol
RUO – Research Use Only