missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic Acid Solution, Honeywell™ Fluka™

Product Code. 15618320
Click to view available options
Quantity:
10 mL
100 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 75-75-2
Molecular Formula CH4O3S
Molecular Weight (g/mol) 96.1
MDL Number MFCD00007518
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
Synonym methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
PubChem CID 6395
ChEBI CHEBI:27376
IUPAC Name methanesulfonic acid
SMILES CS(=O)(=O)O

Specifications

Color Colorless
Physical Form Liquid
Quantity 10 mL
Linear Formula CH3SO3H
UN Number UN3265
Beilstein 1446024
Formula Weight 96.11g/mol
Concentration or Composition (by Analyte or Components) approx. 1M in water
Packaging Glass Bottle
Density 1.039g/mL at 20°C
Chemical Name or Material Methanesulfonic Acid Solution, For ion chromatography
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'