Learn More
Methallyltri-n-butyltin, 98%, Thermo Scientific Chemicals
75.07 € - 276.38 €
Chemical Identifiers
CAS | 67883-62-9 |
---|---|
Molecular Formula | C16H34Sn |
Molecular Weight (g/mol) | 345.158 |
MDL Number | MFCD03425867 |
InChI Key | CYIUFVYZGOCGEV-UHFFFAOYSA-N |
Synonym | methallyltri-n-butyltin, tributyl 2-methylallyl stannane, stannane, tributyl 2-methyl-2-propenyl, tributyl 2-methylprop-2-en-1-yl stannane, methallyltributyltin, 2-methylallyltributyltin, 2-methylallyl tributyltin, methallyltri-n-butylstannane, methallyltri-n-butylin, 2-methylallyl-tributylstannane |
PubChem CID | 11013379 |
IUPAC Name | tributyl(2-methylprop-2-enyl)stannane |
SMILES | CCCC[Sn](CCCC)(CCCC)CC(=C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11319016
|
Thermo Scientific Alfa Aesar
L20186.03 |
1 g |
75.07 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11329016
|
Thermo Scientific Alfa Aesar
L20186.06 |
5 g |
276.38 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used as a methallylating reagent. Also used as a sole curative. It is used as a primary and secondary intermediates in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a methallylating reagent. Also used as a sole curative. It is used as a primary and secondary intermediates in pharmaceuticals.
Solubility
Fully miscible with water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
67883-62-9 | |
345.158 | |
CYIUFVYZGOCGEV-UHFFFAOYSA-N | |
11013379 | |
CCCC[Sn](CCCC)(CCCC)CC(=C)C |
C16H34Sn | |
MFCD03425867 | |
methallyltri-n-butyltin, tributyl 2-methylallyl stannane, stannane, tributyl 2-methyl-2-propenyl, tributyl 2-methylprop-2-en-1-yl stannane, methallyltributyltin, 2-methylallyltributyltin, 2-methylallyl tributyltin, methallyltri-n-butylstannane, methallyltri-n-butylin, 2-methylallyl-tributylstannane | |
tributyl(2-methylprop-2-enyl)stannane |
Specifications
67883-62-9 | |
1.4875 | |
UN2788 | |
Fully miscible with water. | |
CCCC[Sn](CCCC)(CCCC)CC(=C)C | |
345.158 | |
345.16 | |
Methallyltri-n-butyltin |
C16H34Sn | |
MFCD03425867 | |
methallyltri-n-butyltin, tributyl 2-methylallyl stannane, stannane, tributyl 2-methyl-2-propenyl, tributyl 2-methylprop-2-en-1-yl stannane, methallyltributyltin, 2-methylallyltributyltin, 2-methylallyl tributyltin, methallyltri-n-butylstannane, methallyltri-n-butylin, 2-methylallyl-tributylstannane | |
CYIUFVYZGOCGEV-UHFFFAOYSA-N | |
tributyl(2-methylprop-2-enyl)stannane | |
11013379 | |
98% |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.