missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MetFormin hydrochloride, 98%, For HPLC analysis, MP Biomedicals™
41.20 € - 200.00 €
Chemical Identifiers
CAS | 1115-70-4 |
---|---|
Molecular Formula | C4H12ClN5 |
Molecular Weight (g/mol) | 165.63 |
MDL Number | MFCD00012582 |
InChI Key | OETHQSJEHLVLGH-UHFFFAOYSA-N |
Synonym | metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform |
PubChem CID | 14219 |
IUPAC Name | hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride |
SMILES | [H+].[Cl-].CN(C)C(=N)N=C(N)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11462432
|
MP Biomedicals
0215169101 |
1 g |
41.20 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11482432
|
MP Biomedicals
0215169110 |
10 g |
200.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
1115-70-4 | |
165.63 | |
OETHQSJEHLVLGH-UHFFFAOYSA-N | |
14219 | |
[H+].[Cl-].CN(C)C(=N)N=C(N)N |
C4H12ClN5 | |
MFCD00012582 | |
metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform | |
hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride |
Specifications
1115-70-4 | |
C4H12ClN5 | |
metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform | |
[H+].[Cl-].CN(C)C(=N)N=C(N)N | |
165.63 | |
165.6 | |
Crystalline Powder |
White | |
MFCD00012582 | |
OETHQSJEHLVLGH-UHFFFAOYSA-N | |
hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride | |
14219 | |
0.98 | |
Metformin hydrochloride, For HPLC analysis |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Metformin Hydrochloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..