Learn More
Sampling Systems™ Metal Detectable Liquid Funnels
Our plastic Metal Detectable Liquid Funnels are an important part of our metal detectable range. The 100 mm top diameter means they are ideal for most dispensing applications.
Brand: Sampling Systems™ M1023B-100
This item is not returnable.
View return policy
Description
- Our metal detectable funnels have a simple handle near the edge so they are easy to use.
- They are made from a special chemically resistant grade of PP.
- The whole range is manufactured in cleanroom conditions and packed in an ISO class 7 (10,000) cleanroom.
- This ensures the cleanliness of the product.
- The robust design means they can be used for years and years or used as a single use product straight from the bag.
- Simply unwrap and use.
- They are ideal for use with liquids and suspensions.
- Made from high quality PP.
- Conforms to the latest FDA and EU food requirements.
- 100 mm diameter is a standard size.
Specifications
100 mm | |
Polypropylene | |
Blue |