missing translation for 'onlineSavingsMsg'
Learn More

Aroclor 1242 in Methyl-tert.butylether 1000μg/mL, Fisher Chemical™

Product Code. 12916143
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 1000μg/mL
CAS 53469-21-9
Packaging Ampoule
Quantity 1 mL
Type Organic Single-component Standard Solution
Molecular Formula C12H6Cl4
InChI Key ALFHIHDQSYXSGP-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)Cl)Cl)C2=C(C=C(C=C2)Cl)Cl
IUPAC Name 1,2-dichloro-3-(2,4-dichlorophenyl)benzene
Molecular Weight (g/mol) 291.98
PubChem CID 63103
Solvent or Matrix Methyl-tert.butylether
Chemical Name or Material Aroclor 1242
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'