missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobiphenyl in DCM 10μg/mL, Fisher Chemical™

Product Code. 12996773
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule(s)
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 10μg/mL
CAS 321-60-8
Packaging Ampoule(s)
Quantity 10 mL
Molecular Formula C12H9F
MDL Number MFCD00000317
InChI Key KLECYOQFQXJYBC-UHFFFAOYSA-N
SMILES FC1=CC=CC=C1C1=CC=CC=C1
IUPAC Name 2-fluoro-1,1'-biphenyl
Molecular Weight (g/mol) 172.20
PubChem CID 67579
Solvent or Matrix Dichloromethane
Chemical Name or Material 2-Fluorobiphenyl
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'