Learn More
Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 5.0
127.64 € - 195.98 €
Chemical Identifiers
CAS | 145224-94-8 |
---|---|
Molecular Formula | C6H15NO5S |
Molecular Weight (g/mol) | 213.248 |
MDL Number | MFCD00283848 |
InChI Key | MIIIXQJBDGSIKL-UHFFFAOYSA-N |
PubChem CID | 16218417 |
IUPAC Name | 2-morpholin-4-ylethanesulfonic acid;hydrate |
SMILES | C1COCCN1CCS(=O)(=O)O.O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15484439
|
Thermo Scientific Chemicals
J61960.AK |
250 mL |
127.64 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15494439
|
Thermo Scientific Chemicals
J61960.AP |
500 mL |
195.98 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
145224-94-8 | |
213.248 | |
MIIIXQJBDGSIKL-UHFFFAOYSA-N | |
2-morpholin-4-ylethanesulfonic acid;hydrate |
C6H15NO5S | |
MFCD00283848 | |
16218417 | |
C1COCCN1CCS(=O)(=O)O.O |
Specifications
145224-94-8 | |
5.0 | |
C6H15NO5S | |
Miscible with water. | |
C1COCCN1CCS(=O)(=O)O.O | |
213.248 | |
MES buffer soln. |
1.0 M | |
Liquid | |
MFCD00283848 | |
MIIIXQJBDGSIKL-UHFFFAOYSA-N | |
2-morpholin-4-ylethanesulfonic acid;hydrate | |
16218417 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.