Learn More
Mercury(II) thiocyanate, 99+%, Thermo Scientific Chemicals
74.80 € - 305.00 €
Chemical Identifiers
CAS | 592-85-8 |
---|---|
Molecular Formula | C2HgN2S2 |
Molecular Weight (g/mol) | 316.75 |
MDL Number | MFCD00011039 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10296440
|
Thermo Scientific Acros
196890500 |
50 g | Glass bottle |
74.80 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15730829
|
Thermo Scientific Acros
196892500 |
250 g | Glass bottle |
305.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
592-85-8 | |
C2HgN2S2 | |
316.75 | |
MFCD00011039 |
Specifications
165.0°C | |
White | |
99% min. (Complexometry) | |
Hg(SCN)2 | |
09,293 | |
15, 5958 | |
316.75 | |
≥99% |
592-85-8 | |
Crystalline Powder | |
C2HgN2S2 | |
MFCD00011039 | |
120°C | |
Solubility in water: 0.7g/L (25°C). Other solubilities: soluble in diluted hcl | |
316.75 | |
Mercury(II) thiocyanate |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at r
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-773-
- Mercury(II) thiocyanate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P330-Rinse mouth.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only