missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) sulfate, 98+%, Thermo Scientific Chemicals

Product Code. 11407807
Click to view available options
Quantity:
50 g
250 g
1000 g

It is used in medicine as a catalyst and in extracting gold and silver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine as a catalyst and in extracting gold and silver.

Solubility
It is soluble in hot H2SO4, NaCl solution. insoluble in alcohol, acetone, ammonia

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Light sensitive. Moisture sensitive. Air sensitive. Keep away from Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Melting Point (decomposition)
Quantity 1000 g
Linear Formula HgSO4
UN Number UN1645
Merck Index 14,5887
Solubility Information It is soluble in hot H2SO4,NaCl solution. insoluble in alcohol,acetone,ammonia
IUPAC Name mercury(2+);sulfate
Formula Weight 296.65
Percent Purity ≥98%
Sensitivity Moisture sensitive
Density 6.47
Chemical Name or Material Mercury(II) sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'