Learn More
Mercury(II) perchlorate trihydrate, Reagent Grade, Thermo Scientific™
75.00 € - 201.00 €
Chemical Identifiers
CAS | 73491-34-6 |
---|---|
Molecular Formula | Cl2H6HgO11 |
Molecular Weight (g/mol) | 453.529 |
MDL Number | MFCD00149735 |
InChI Key | DFJWPGPUWHLJIJ-UHFFFAOYSA-L |
Synonym | mercury ii perchlorate trihydrate, mercury perchlorate-water 1/2/3, perchloric acid,mercury 2+ salt, trihydrate 9ci |
PubChem CID | 53471339 |
IUPAC Name | mercury(2+);diperchlorate;trihydrate |
SMILES | O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Hg+2] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
73491-34-6 | |
453.529 | |
DFJWPGPUWHLJIJ-UHFFFAOYSA-L | |
53471339 | |
O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Hg+2] |
Cl2H6HgO11 | |
MFCD00149735 | |
mercury ii perchlorate trihydrate, mercury perchlorate-water 1/2/3, perchloric acid,mercury 2+ salt, trihydrate 9ci | |
mercury(2+);diperchlorate;trihydrate |
Specifications
73491-34-6 | |
Crystalline | |
Hg(ClO4)2·3H2O | |
UN3087 | |
Soluble in water. Insoluble in benzene and hexane. | |
O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Hg+2] | |
453.529 | |
453.54 (399.49 Anhydrous) | |
Mercury(II) perchlorate trihydrate |
White | |
Cl2H6HgO11 | |
MFCD00149735 | |
mercury ii perchlorate trihydrate, mercury perchlorate-water 1/2/3, perchloric acid,mercury 2+ salt, trihydrate 9ci | |
DFJWPGPUWHLJIJ-UHFFFAOYSA-L | |
mercury(2+);diperchlorate;trihydrate | |
53471339 | |
Reagent |
Safety and Handling
GHS H Statement
H271-H300-H310-H330-H373
May cause fire or explosion
strong oxidizer.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P210-P220-P221-P260-P262-P264b-P270-P271-P280-P283-P284-P301+P310-P302+P350-P304+P340-P306+P360-P310-P314-P330-P363-P370+P378q-P371+P380+P375-P501c
H271-H300+H310+H330-H373
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 231-525-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.