Learn More
Mercury(II) oxide, yellow, ACS reagent, Thermo Scientific Chemicals
62.60 € - 218.00 €
Chemical Identifiers
CAS | 21908-53-2 |
---|---|
Molecular Formula | HgO |
Molecular Weight (g/mol) | 216.59 |
MDL Number | MFCD00011045 |
InChI Key | UKWHYYKOEPRTIC-UHFFFAOYSA-N |
Synonym | mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide |
IUPAC Name | oxomercury |
SMILES | O=[Hg] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10237520
|
Thermo Scientific Acros
316791000 |
100 g | Glass bottle |
62.60 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10178093
|
Thermo Scientific Acros
316795000 |
500 g | Glass bottle |
218.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
21908-53-2 | |
216.59 | |
UKWHYYKOEPRTIC-UHFFFAOYSA-N | |
oxomercury |
HgO | |
MFCD00011045 | |
mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide | |
O=[Hg] |
Specifications
500.0°C | |
Yellow to Orange | |
HgO | |
MFCD00011045 | |
12, 5936 | |
UKWHYYKOEPRTIC-UHFFFAOYSA-N | |
oxomercury | |
216.59 | |
ACS Reagent |
21908-53-2 | |
Powder | |
HgO | |
01,655; 02,267; 04,323; 05,428; 06,360; 07,224; 08,316; 09,293; 11,318; 12,305; 14,204; 15,174 | |
mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide | |
O=[Hg] | |
216.59 | |
≥99.0% | |
Mercury(II) oxide, yellow |
Safety and Handling
GHS H Statement
Fatal in contact with skin.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
Fatal if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 244-654-7
- Mercury(II) oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H310-Fatal in contact with skin.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H300-Fatal if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P361-Take off immediately all contaminated clothing.
RUO – Research Use Only