missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(II) Iodide, Extra Pure, SLR, Fisher Chemical™
81.45 € - 642.00 €
Chemical Identifiers
CAS | 7774-29-0 |
---|---|
Molecular Formula | HgI2 |
Molecular Weight (g/mol) | 454.401 |
MDL Number | 11044 |
InChI Key | YFDLHELOZYVNJE-UHFFFAOYSA-L |
Synonym | mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum |
PubChem CID | 24485 |
ChEBI | CHEBI:49659 |
IUPAC Name | diiodomercury |
SMILES | I[Hg]I |
Description
Chemical Identifiers
7774-29-0 | |
454.401 | |
YFDLHELOZYVNJE-UHFFFAOYSA-L | |
24485 | |
diiodomercury |
HgI2 | |
11044 | |
mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum | |
CHEBI:49659 | |
I[Hg]I |
Specifications
354°C | |
7774-29-0 | |
8 | |
HgI2 | |
mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum | |
I[Hg]I | |
454.401 | |
CHEBI:49659 | |
Plastic powder jar |
259°C | |
Red | |
Solid | |
11044 | |
YFDLHELOZYVNJE-UHFFFAOYSA-L | |
diiodomercury | |
24485 | |
454.39g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Mercury(II) iodide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title