missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Acetate, 99.99% Trace Metals Basis, Honeywell™

Product Code. 15625570
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

99.999% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1600-27-7
Molecular Formula C4H6HgO4
Molecular Weight (g/mol) 318.68
MDL Number MFCD00012165
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
Synonym mercury 2+ ion acetic acid
PubChem CID 15337
ChEBI CHEBI:33211
IUPAC Name mercury(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Hg+2]

Specifications

Melting Point 179°C to 182°C
Color Beige
Quantity 100 g
Linear Formula (CH3COO)2Hg
UN Number UN1629
Beilstein 3563831
Formula Weight 318.68g/mol
Percent Purity 99.999% (trace metals basis)
Packaging Poly Bottle
Appearance Powder
Density 3.27g/cm3
Chemical Name or Material Mercury(II) acetate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'