missing translation for 'onlineSavingsMsg'
Learn More

Mercury(I) nitrate dihydrate, 97%, Thermo Scientific Chemicals

Product Code. 11417737
Click to view available options
Quantity:
50 g
250 g
1000 g

Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with alcohols, sulfur and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14836-60-3
Molecular Formula Hg2(NO3)2·2H2O
MDL Number MFCD00150812
Synonym Mercurous nitrate

Specifications

Melting Point 70°C
Quantity 250 g
Linear Formula Hg2(NO3)2·2H2O
UN Number UN1627
Merck Index 14,5896
Solubility Information Slightly soluble in water.
Formula Weight 561.22 (525.19 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Light sensitive
Density 4.79
Chemical Name or Material Mercury(I) nitrate dihydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.