missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Menzel™ Microscope Coverslips

Brand:  Epredia™ BB02400600A113FST0

Product Code. 15747592

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Coverslip
Borosilicate Glass
60 mm
24 x 60 mm
1000/Pk.
Square
24 mm
#1 (0.13 to 0.16 mm)
Automated equipment
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ Menzel™ Microscope Coverslips > Length: 24mm; Width: 60mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.