Learn More
Serva Electrophoresis™ Membra-Cel™ Dialysis Tubing
Specifications
Product Type Dialysis Tubing
MWCO 3.5 kDa
Material Cellulose
Diameter (Metric) 16 mm
Capacity (Metric) 2 mL/cm
View More Specs
Specifications
Dialysis Tubing
Cellulose
2 mL/cm
3.5 kDa
16 mm
1 Each
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Serva Electrophoresis™ Membra-Cel™ Dialysis Tubing