missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melting Point (Benzophenone, Benzoic Acid, Anthraquinone) Standard Set, Reagecon
Specially prepared to calibrate, control, verify or qualify melting point (MP) devices.
Brand: Reagecon RMPSET2
This item is not returnable.
View return policy
Description
- Melting Point Reference Standard, Melting Point Reference Material
- The exceptional purity of this product is critical as MP will change with the presence of very small quantities of impurities.
- This product is produced in accordance with the European, US and Chinese Pharmacopeia requirements.
- The product can assist in identifying a compound or derivative or checking the purity or thermal stability of a known compound.
- Tested and certified using a reference melting point apparatus that is calibrated using Certified Reference Materials to give traceability to the ITS-90 Temperature Scale.
- Both the meniscus formation and complete liquefaction temperatures are certified.
Specifications
Accuracy ± 0.03°C | |
3 x 1 g | |
Solid |
Blue Glass Bottle | |
Benzophenone 47 to 49°C Standard, Benzoic Acid 121 to 123°C Standard, Anthraquinone 283 to 286°C Standard |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- X3 MELTING P. STD BENZOPHENONE,BENZ,ANTHRAQUI10GR
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : anthraquinone, benzoic acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction