missing translation for 'onlineSavingsMsg'
Learn More

Melting Point Benzophenone 47 to 49°C Standard, Reagecon
BIOPHARMA

Ultrapure substance, specially prepared to calibrate, control, verify or qualify melting point (MP) devices.

Brand:  Reagecon RMP048

Product Code. 13572100

  • 16.55 € / 1g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Melting Point Reference Standard, Melting Point Reference Material
  • The exceptional purity of this product is critical as MP will change with the presence of very small quantities of impurities.
  • This product is produced in accordance with the European, US and Chinese Pharmacopeia requirements.
  • The product can assist in identifying a compound or derivative or checking the purity or thermal stability of a known compound.
  • Tested and certified using a reference melting point apparatus that is calibrated using Certified Reference Materials to give traceability to the ITS-90 Temperature Scale.
  • Both the meniscus formation and complete liquefaction temperatures are certified.
Specifications

Specifications

Accuracy ± 0.03°C
1 x 1 g
Solid
Blue Glass Bottle
Benzophenone 47 to 49°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.