Learn More
DWK Life Sciences Wheaton™ Media Lab Bottle, Non-Graduated, Capless
Product Code. 12442501
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
Quantity:
24 EA
48 EA
Clear Type I borosilicate glass bottle without graduations.
  • Manufactured from borosilicate glass which provides the best chemical resistance
  • Clear borosilicate glass bottle
  • Special pour lip facilitates pouring
  • Packaged in partitioned carton to help prevent scratch marks on bottles
  • Black phenolic screw cap can be purchased separately
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Color Clear
Material Borosilicate Glass (Type I)
Quantity 48 EA
Neck Finish 33-340
Product Type Media Bottle
Closure Size 33-340 mm
Closure Type No Closure