missing translation for 'onlineSavingsMsg'
Learn More

VACUUBRAND™ ME 4C NT +2AK Chemistry Vacuum System

Use this pump system for evacuating and pumping large amounts of gases and vapors. Vacubrand™ ME 4C NT +2AK Chemistry Vacuum Systems provide a combination of high pumping speed and low ultimate vacuum down to 70 mbar.

Brand:  VACUUBRAND™ 22614080

2981.33 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 11385670

  • 3320.00 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Chemistry vacuum system with a wide range of applications in chemical, biological and pharmaceutical laboratories if condensation of solvent vapors at the outlet is not necessary
  • Typical applications are vacuum ovens, single and multiple filtrations, as well as concentration of solvents with low boiling points
  • Separator at the inlet (AK) is made of glass with protective coating and retains particles and liquid droplets
  • Separator at the outlet collects condensate, avoids condensate backflow towards the pump and makes the pump whisper-quiet
  • Outstanding chemical resistance and superior vapor tolerance
  • High performance even at low vacuum levels
  • Compact design
  • Whisper quiet and very low vibration
  • Separators at inlet and outlet to collect condensates

Order Info

Includes

TRUSTED_SUSTAINABILITY
Specifications

Specifications

52.5 torr
IEC Plug EN 60320, Dual Voltage Motor, Manual
ATEX Conformity II 3G IIC T3 X, CAN/CSA C22.2 61010-1-12, CE, DIN EN 1012-2:2011, DIN EN 12100:2001, DIN EN 50581:2013, DIN EN 61010-1:2011, DIN EN 61326-1:2013, IEC 61010-1:2010, NRTL Certification, TUV, UL 61010A 61010-1:2012
316 x 242 x 291 mm
ME 4C NT +2AK
-10°C to +60°C
40°C
2.3/2.6 CFM
292.1 mm
243.84 mm
IP40
Chemical-Resistant
Concentration of Solvents with Low Boiling Points
1
100 to 115/120/200 to 230 V
0.24 hp
150 to 1800 rpm
230 V 50/60 Hz
Fluoroplastic (Case)
45 dB(A)
104°F
10°C
50/60 Hz
314.96 mm
Diaphragm Pump
2
50°F
Oil-Free
16 psi
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.