missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15462-1-AP-150UL
This item is not returnable.
View return policy
Description
MDH2 is localized to the mitochondria and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.Specifications
MDH2 | |
Polyclonal | |
Unconjugated | |
MDH2 | |
M MDH, MDH, MDH2, MOR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17448, 4191, 81829 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P04636, P08249, P40926 | |
Mdh2 | |
MDH2 Fusion Protein Ag7750 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction