missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10513-1-AP-150UL
This item is not returnable.
View return policy
Description
MCM2 (Mini chromosome maintece protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. MCM2 has been mapped to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).Specifications
MCM2 | |
Polyclonal | |
Unconjugated | |
MCM2 | |
BM28, CCNL1, cdc19, CDCL1, D3S3194, KIAA0030, MCM2, MITOTIN, Nuclear protein BM28 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17216, 312538, 4171 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49736, P97310 | |
Mcm2 | |
MCM2 Fusion Protein Ag0798 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction