missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAZ Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21068-1-AP-20UL
This item is not returnable.
View return policy
Description
May function as a transcription factor with dual roles in transcription initiation and termination. Binds to two sites, ME1a1 and ME1a2, within the MYC promoter having greater affinity for the former. Also binds to multiple G/C-rich sites within the promoter of the Sp1 family of transcription factors.Specifications
MAZ | |
Polyclonal | |
Unconjugated | |
MAZ | |
MAZ, MAZI, Pur 1, PUR1, SAF 1, SAF 2, ZF87, Zif87, ZNF801 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4150 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P56270 | |
MAZ | |
MAZ Fusion Protein Ag15200 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction