Learn More
MAZ Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21068-1-AP-150UL
 View more versions of this product
Product Code. 16800675
  • 410.00 € / 150µL
Explore more special offers
Description
Description
May function as a transcription factor with dual roles in transcription initiation and termination. Binds to two sites, ME1a1 and ME1a2, within the MYC promoter having greater affinity for the former. Also binds to multiple G/C-rich sites within the promoter of the Sp1 family of transcription factors.
Specifications
Specifications
MAZ
Polyclonal
Unconjugated
MAZ
MAZ, MAZI, Pur 1, PUR1, SAF 1, SAF 2, ZF87, Zif87, ZNF801
Rabbit
Antigen Affinity Chromatography
RUO
4150
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P56270
MAZ
MAZ Fusion Protein Ag15200
150 μL