Learn More
3M™ Maxim™ DX Safety Glasses
Click to view available options
Lens Tint:
Clear
Shade 3
Shade 5
Frame Color:
Black, Gray
Blue, Gray
Description
- Safety spectacles with aspherical lens for 180° of Pure Vision and superior eye coverage
- DX coating provides enhanced anti-scratch and anti-fog protection
- Spectacles with ventilation channel across the lens to minimize misting
- Design provides excellent coverage and good field of vision
- Pliable, adjustable nose-pads and a 'soft-touch' brow guard
- Adjustable temple lengths and pantoscopic (angle) adjustments
Specifications
Specifications
Certifications/Compliance | EN 166: EN 169 |
Lens Type | DX |
Lens Tint | Shade 5 |
Product Type | Safety glasses |
For Use With (Application) | Construction |
Frame Color | Black, Gray |
Lens Material | Polycarbonate |