missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ L/S™ Easy-Load™ 3 Pump Head
Mount and stack these pump heads without tools or hardware. Automatic tubing retention speeds setup and changeover. Masterflex™ L/S™ Easy-Load™ 3 Pump Heads feature an automatic retention mechanism that makes loading and unloading tubing easier than ever.
Brand: Masterflex™ 77800-60
489.10 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
- Flow range: 0.06 to 2300mL/min depending on drive rpm and tubing size
- Each pump head for precision pump tubing accepts all six precision tubing sizes: L/S 13, L/S 14, L/S 16, L/S 25, L/S 17, and L/S 18
- Twist-lock mounting feature lets you mount and stack heads in seconds.
- Included mounting plate adapts Easy-Load 3 to L/S drives
- Locking tab secures pump head during operation in either direction
- Polypropylene (PP) housing with CRS or SS rotor
- Same side tubing entry/exit permits easy integration into space-limited applications
- Mount and operate pump head in any of four positions (depending on drive)
- Stack up to four heads to increase flow capacity and for multiple-channel applications
Specifications
For precision tubing | |
35 to 36mm | |
0.06 to 2300mL/min. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Masterflex™ L/S™ Easy-Load™ 3 Pump Head > Length Metric: 15m
Spot an opportunity for improvement?Share a Content Correction