Learn More
Atago™ MASTER-S/MillM Model Salinity Refractometer
MASTER-S/MillM Model Salinity Refractometer
Brand:  Atago™ 2493
Product Code. 11486148
  • 335.00 € / Each
Explore more special offers
Description
Description
MASTER-S/MillM Model Salinity Refractometer
MASTER-S/MillM Model Salinity Refractometer
Specifications
Specifications
Salinity Refractometer
MASTER-S/MillM
Measures the salinity and specific gravity of seawater, and utilizes a new material that is durable against salty samples
0 to 100 Salinity, 1.000 to 1.070 Specific Gravity
Salinity, Specific gravity
110 g