Learn More
Atago™ MASTER-S/Millα Salinity Refractometer
Ideal for measuring the salinity and specific gravity of seawater. Atago™ MASTER-S/Millα Salinity Refractometer uses a new material that is durable against salty samples.
Brand:  Atago™ 2491
Product Code. 11476148
  • 377.00 € / Each
Explore more special offers
Description
Description
  • Durable against salty samples
  • Displays salinity in parts per million
  • Automatic temperature compensation
  • Water resistant
Specifications
Specifications
Salinity Refractometer
MASTER-S/Millα
±1‰ Salinity, ±0.0005 Specific gravity
Measuring the salinity and specific gravity of seawater
110 g
0 to 100 Salinity, 1.000 to 1.070 Specific Gravity
±2‰ Salinity, ±0.001 Specific gravity at 10 to 30°C
Salinity, Specific gravity
IP65
3.2 x 3.4 x 20.7 cm