missing translation for 'onlineSavingsMsg'
Learn More

Marble Chips, Technical, Granule Size 20-30mm, Fisher Chemical™

Product Code. 11408982
Click to view available options
Quantity:
1 kg
3 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 825°C
Color White
pH 8 to 9
Physical Form Solid
Quantity 1 kg
Formula Weight 100.09g/mol
Packaging Plastic powder jar
Product Identifier
  • MARBLE CHIPS 2-3MM
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'