missing translation for 'onlineSavingsMsg'
Learn More

Manganous Chloride, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15518024
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 197.9
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.01kg
Chemical Name or Material Manganous Chloride, 4-Hydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese (II) chloride tetrahydrate, Crystal BAKER ANALYZED A.C.S. Reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Manganese (II) chloride tetrahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'