missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese powder, -325 mesh, 99.3% (metals basis), Thermo Scientific Chemicals
Form: Apparent density 2.6-3.5g/cm3
37.99 € - 188.94 €
Chemical Identifiers
CAS | 7439-96-5 |
---|---|
Molecular Formula | Mn |
Molecular Weight (g/mol) | 54.94 |
MDL Number | MFCD00011111 |
InChI Key | PWHULOQIROXLJO-UHFFFAOYSA-N |
Synonym | colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds |
PubChem CID | 23930 |
ChEBI | CHEBI:35154 |
IUPAC Name | manganese |
SMILES | [Mn] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11301308
|
Thermo Scientific Alfa Aesar
010238.14 |
25 g |
37.99 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381298
|
Thermo Scientific Alfa Aesar
010238.30 |
250 g |
66.33 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391298
|
Thermo Scientific Alfa Aesar
010238.A1 |
1 kg |
188.94 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-96-5 | |
54.94 | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
23930 | |
manganese |
Mn | |
MFCD00011111 | |
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds | |
CHEBI:35154 | |
[Mn] |
Specifications
Gray | |
Gray powder | |
Mn | |
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds | |
[Mn] | |
54.94 | |
CHEBI:35154 | |
Powder |
7439-96-5 | |
99.3% (Metals basis) | |
MFCD00011111 | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
manganese | |
23930 | |
-325 Mesh | |
Manganese |
Safety and Handling
P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
H228-H319
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III
missing translation for 'einecsNumber' : 231-105-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only