Learn More
Manganese(IV) oxide activated, tech. 90%, Thermo Scientific Chemicals
23.80 € - 66.90 €
Chemical Identifiers
CAS | 1313-13-9 |
---|---|
Molecular Formula | MnO2 |
Molecular Weight (g/mol) | 86.94 |
MDL Number | MFCD00003463 |
InChI Key | NUJOXMJBOLGQSY-UHFFFAOYSA-N |
Synonym | manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein |
PubChem CID | 14801 |
IUPAC Name | dioxomanganese |
SMILES | O=[Mn]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468979
|
Thermo Scientific Alfa Aesar
L13253.09 |
10 g |
23.80 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15478979
|
Thermo Scientific Alfa Aesar
L13253.22 |
100 g |
29.80 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15488979
|
Thermo Scientific Alfa Aesar
L13253.36 |
500 g |
66.90 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Manganese(IV) oxide is suitable for use in batteries. It is widely used for the selective oxidation of allylic and benzylic alcohols to aldehydes or ketones, as a co-oxidant in combination with 1,4-benzoquinone and in the allylic oxidation of olefins catalyzed by Palladium(II) acetate. It acts a reagent for oxidations. It is the source of manganese and all its compounds, largely used in manufacturing of manganese steel, for making amethyst glass, decolorizing glass, painting on porcelain, faience and majolica. The precipitate is used in electrotechnics, pigments, browning gun barrels, drier for paints and varnishes, printing and dyeing textiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsManganese(IV) oxide is suitable for use in batteries. It is widely used for the selective oxidation of allylic and benzylic alcohols to aldehydes or ketones, as a co-oxidant in combination with 1,4-benzoquinone and in the allylic oxidation of olefins catalyzed by Palladium(II) acetate. It acts a reagent for oxidations. It is the source of manganese and all its compounds, largely used in manufacturing of manganese steel, for making amethyst glass, decolorizing glass, painting on porcelain, faience and majolica. The precipitate is used in electrotechnics, pigments, browning gun barrels, drier for paints and varnishes, printing and dyeing textiles.
Solubility
Insoluble in water.
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
1313-13-9 | |
86.94 | |
NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
14801 | |
O=[Mn]=O |
MnO2 | |
MFCD00003463 | |
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
dioxomanganese |
Specifications
∽535°C (decomposition) | |
MnO2 | |
14,5730 | |
Insoluble in water. | |
O=[Mn]=O | |
86.94 | |
86.94 | |
Technical | |
Manganese(IV) oxide activated |
1313-13-9 | |
MFCD00003463 | |
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
dioxomanganese | |
14801 | |
90% | |
5.026 |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
H302+H332-H373
missing translation for 'einecsNumber' : 215-202-6
missing translation for 'rtecsNumber' : OP0350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only