missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) Sulfate Tetrahydrate, 98+%, Extra Pure, SLR, Fisher Chemical™
138.00 € - 161.00 €
Chemical Identifiers
CAS | 10034-96-5 |
---|---|
Molecular Formula | H2MnO5S |
Molecular Weight (g/mol) | 169.01 |
MDL Number | MFCD00149159 |
InChI Key | ISPYRSDWRDQNSW-UHFFFAOYSA-L |
Synonym | manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate |
PubChem CID | 177577 |
ChEBI | CHEBI:86364 |
IUPAC Name | manganese(2+);sulfate;hydrate |
SMILES | O.[Mn++].[O-]S([O-])(=O)=O |
Description
Manganese(II) Sulfate Tetrahydrate, 98+%, Extra Pure, SLR
Chemical Identifiers
10034-96-5 | |
169.01 | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
177577 | |
manganese(2+);sulfate;hydrate |
H2MnO5S | |
MFCD00149159 | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
CHEBI:86364 | |
O.[Mn++].[O-]S([O-])(=O)=O |
Specifications
26°C to 27°C | |
Red | |
H2MnO5S | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
O.[Mn++].[O-]S([O-])(=O)=O | |
169.01 | |
CHEBI:86364 | |
Plastic powder jar |
10034-96-5 | |
Solid | |
MFCD00149159 | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
manganese(2+);sulfate;hydrate | |
177577 | |
223.05g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Manganese(II) sulfate tetrahydrate
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H318-Causes serious eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title