missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) nitrate hexahydrate, 98+%, Thermo Scientific Chemicals
24.40 € - 118.00 €
Specifications
Melting Point | ca 26°C |
---|---|
CAS | 17141-63-8 |
Physical Form | Solid |
Molecular Formula | MnN{2}O{6}^0.6 H{2}O |
Percent Purity | ≥98% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11328925
|
Thermo Scientific Alfa Aesar
L14040.14 |
25 g |
24.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11338925
|
Thermo Scientific Alfa Aesar
L14040.22 |
100 g |
47.10 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348925
|
Thermo Scientific Alfa Aesar
L14040.36 |
500 g |
118.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
ca 26°C | |
Solid | |
≥98% |
17141-63-8 | |
MnN{2}O{6}^0.6 H{2}O |